Assay Details
| Gene Symbol: Sema3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042779.2 | NP_001036244.1 | 5 - 6 | 671 | 73 | |
| NM_001291537.1 | NP_001278466.1 | 5 - 6 | 935 | 73 | ||
| NM_001291538.1 | NP_001278467.1 | 5 - 6 | 929 | 73 | ||
| NM_001291539.1 | NP_001278468.1 | 4 - 5 | 612 | 73 | ||
| NM_009153.3 | NP_033179.2 | 3 - 4 | 481 | 73 | ||
| NR_111986.1 | 3 - 4 | 491 | 73 | |||
| GenBank mRNA | AK014333.1 | - | 3 - 4 | 427 | 73 | |
| AK033597.1 | - | 4 - 5 | 622 | 73 | ||
| AK037034.1 | - | 5 - 6 | 660 | 73 | ||
| AK043634.1 | - | 4 - 5 | 565 | 73 | ||
| AK048364.1 | - | 4 - 5 | 893 | 73 | ||
| BC090669.1 | - | 4 - 5 | 592 | 30696359 | 73 | |
| BX515696.1 | - | 2 - 3 | 330 | 73 | ||
| X85990.1 | - | 3 - 4 | 481 | 73 | ||
More Information
Set Membership: |
|

Sema3b
Genomic Map