Assay Details
| Gene Symbol: Sema3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042779.2 | NP_001036244.1 | 9 - 10 | 1063 | 65 | |
| NM_001291537.1 | NP_001278466.1 | 9 - 10 | 1327 | 65 | ||
| NM_001291538.1 | NP_001278467.1 | 9 - 10 | 1321 | 65 | ||
| NM_001291539.1 | NP_001278468.1 | 8 - 9 | 1004 | 65 | ||
| NM_009153.3 | NP_033179.2 | 7 - 8 | 873 | 65 | ||
| NR_111986.1 | 7 - 8 | 883 | 65 | |||
| GenBank mRNA | AK014333.1 | - | 7 - 8 | 819 | 65 | |
| AK033597.1 | - | 8 - 9 | 1014 | 65 | ||
| AK037034.1 | - | 9 - 10 | 1052 | 65 | ||
| AK043634.1 | - | 8 - 9 | 957 | 65 | ||
| AK048364.1 | - | 8 - 9 | 1285 | 65 | ||
| AK148232.1 | - | 9 - 10 | 1024 | 65 | ||
| BC090669.1 | - | 8 - 9 | 984 | 30696359 | 65 | |
| BC150758.1 | - | 6 - 7 | 719 | 9087669 | 65 | |
| CN539668.1 | - | 4 - 5 | 406 | 65 | ||
| X85990.1 | - | 7 - 8 | 870 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema3b
Genomic Map