Assay Details
| Gene Symbol: Mapk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081567.1 | NP_001075036.1 | 2 - 3 | 555 | 86 | |
| NM_009158.2 | NP_033184.2 | 2 - 3 | 555 | 86 | ||
| GenBank mRNA | AK036275.1 | - | 2 - 3 | 480 | 86 | |
| AK082242.1 | - | 2 - 3 | 558 | 86 | ||
| AK163206.1 | - | 2 - 3 | 506 | 86 | ||
| BC026697.1 | - | 2 - 3 | 520 | 86 | ||
| BC046625.1 | - | 2 - 3 | 494 | 6494163 | 86 | |
| L35236.1 | - | 2 - 3 | 298 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK163206.1,BC046625.1,L35236.1,NM_001081567.1,NM_009158.2 transcripts.
Set Membership: |
|

Mapk10
Genomic Map