Assay Details
| Gene Symbol: Slc1a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009201.2 | NP_033227.2 | 2 - 3 | 1168 | 99 | |
| GenBank mRNA | AF246130.1 | - | 2 - 3 | 613 | 99 | |
| AK148346.1 | - | 2 - 3 | 1168 | 99 | ||
| AK157838.1 | - | 2 - 3 | 1147 | 99 | ||
| BC037462.1 | - | 2 - 3 | 759 | 4192790 | 99 | |
| D85044.1 | - | 2 - 3 | 1101 | 99 | ||
| L42115.1 | - | 2 - 3 | 1144 | 99 | ||
More Information
Set Membership: |
|

Slc1a5
Genomic Map