Assay Details
| Gene Symbol: Slc4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009208.3 | NP_033234.2 | 15 - 16 | 2558 | 72 | |
| GenBank mRNA | AK147218.1 | - | 2895 | 72 | ||
| BC066086.1 | - | 16 - 17 | 2432 | 72 | ||
| BC080271.1 | - | 15 - 16 | 2553 | 6831942 | 72 | |
| M28383.1 | - | 15 - 16 | 2528 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a3
Genomic Map