Assay Details
| Gene Symbol: Stxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113569.1 | NP_001107041.1 | 9 - 10 | 943 | 61 | |
| NM_009295.2 | NP_033321.2 | 9 - 10 | 938 | 61 | ||
| GenBank mRNA | AF326545.1 | - | 9 - 10 | 797 | 61 | |
| AF326563.1 | - | 9 - 10 | 797 | 61 | ||
| AK004780.1 | - | 4 - 5 | 662 | 61 | ||
| BC031728.1 | - | 9 - 10 | 870 | 4480978 | 61 | |
| D45903.1 | - | 9 - 10 | 832 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stxbp1
Genomic Map