Assay Details
| Gene Symbol: Syp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009305.2 | NP_033331.2 | 4 - 5 | 538 | 72 | |
| GenBank mRNA | AK034291.1 | - | 4 - 5 | 481 | 72 | |
| AK043756.1 | - | 4 - 5 | 435 | 72 | ||
| AK090259.1 | - | 4 - 5 | 436 | 72 | ||
| BC014823.1 | - | 4 - 5 | 408 | 4507116 | 72 | |
| BC052354.1 | - | 4 - 5 | 432 | 72 | ||
| BC115644.1 | - | 3 - 4 | 391 | 72 | ||
| BC115645.1 | - | 3 - 4 | 390 | 72 | ||
| X95818.1 | - | 4 - 5 | 407 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syp
Genomic Map