Assay Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009320.4 | NP_033346.2 | 4 - 5 | 659 | 69 | |
| GenBank mRNA | AK041141.1 | - | 4 - 5 | 632 | 69 | |
| AK143510.1 | - | 4 - 5 | 623 | 69 | ||
| AK162776.1 | - | 4 - 5 | 623 | 69 | ||
| BB636410.1 | - | 4 - 5 | 632 | 69 | ||
| BC015245.1 | - | 4 - 5 | 596 | 4242193 | 69 | |
| L03292.2 | - | 4 - 5 | 631 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a6
Genomic Map