Assay Details
| Gene Symbol: Tgfbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009371.3 | NP_033397.3 | 5 - 6 | 1657 | 92 | |
| NM_029575.3 | NP_083851.3 | 4 - 5 | 1582 | 92 | ||
| GenBank mRNA | AF406755.1 | - | 4 - 5 | 1256 | 92 | |
| AK043619.1 | - | 5 - 6 | 1741 | 92 | ||
| AK046561.1 | - | 5 - 6 | 1583 | 92 | ||
| AK053793.1 | - | 5 - 6 | 1658 | 92 | ||
| AK148167.1 | - | 4 - 5 | 1582 | 92 | ||
| AK159327.1 | - | 5 - 6 | 1637 | 92 | ||
| BC052629.1 | - | 1559 | 92 | |||
More Information
Set Membership: |
|

Tgfbr2
Genomic Map