Assay Details
| Gene Symbol: Tgfbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009371.3 | NP_033397.3 | 6 - 7 | 1792 | 86 | |
| NM_029575.3 | NP_083851.3 | 5 - 6 | 1717 | 86 | ||
| GenBank mRNA | AF406755.1 | - | 5 - 6 | 1391 | 86 | |
| AK043619.1 | - | 6 - 7 | 1876 | 86 | ||
| AK046561.1 | - | 6 - 7 | 1718 | 86 | ||
| AK053793.1 | - | 6 - 7 | 1793 | 86 | ||
| AK148167.1 | - | 5 - 6 | 1717 | 86 | ||
| AK159327.1 | - | 6 - 7 | 1772 | 86 | ||
| BC052629.1 | - | 1694 | 86 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbr2
Genomic Map