Assay Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190974.1 | NP_001177903.1 | 11 - 12 | 1747 | 86 | |
| NM_001190975.1 | NP_001177904.1 | 10 - 11 | 1483 | 86 | ||
| NM_009465.4 | NP_033491.2 | 12 - 13 | 1774 | 86 | ||
| GenBank mRNA | AK033237.1 | - | 11 - 12 | 1716 | 86 | |
| AK155567.1 | - | 13 - 14 | 1660 | 86 | ||
| AK161294.1 | - | 5 - 6 | 447 | 86 | ||
| AK170010.1 | - | 11 - 12 | 1721 | 86 | ||
| AK170897.1 | - | 11 - 12 | 1380 | 86 | ||
| BC008551.1 | - | 7 - 8 | 864 | 86 | ||
| BC046618.1 | - | 12 - 13 | 1611 | 6494383 | 86 | |
| BC050914.1 | - | 12 - 13 | 1582 | 6418252 | 86 | |
| BC058230.1 | - | 12 - 13 | 1635 | 6313662 | 86 | |
| X63535.1 | - | 12 - 13 | 1523 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Axl
Genomic Map