Assay Details
| Gene Symbol: Axl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190974.1 | NP_001177903.1 | 15 - 16 | 2135 | 87 | |
| NM_001190975.1 | NP_001177904.1 | 14 - 15 | 1871 | 87 | ||
| NM_009465.4 | NP_033491.2 | 16 - 17 | 2162 | 87 | ||
| GenBank mRNA | AK155567.1 | - | 17 - 18 | 2048 | 87 | |
| AK161294.1 | - | 9 - 10 | 835 | 87 | ||
| AK170010.1 | - | 15 - 16 | 2109 | 87 | ||
| AK170897.1 | - | 15 - 16 | 1770 | 87 | ||
| BC008551.1 | - | 11 - 12 | 1252 | 87 | ||
| BC046618.1 | - | 16 - 17 | 1999 | 6494383 | 87 | |
| BC050914.1 | - | 16 - 17 | 1970 | 6418252 | 87 | |
| BC058230.1 | - | 16 - 17 | 2023 | 6313662 | 87 | |
| X63535.1 | - | 16 - 17 | 1911 | 87 | ||
More Information
Set Membership: |
|

Axl
Genomic Map