Assay Details
| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009604.3 | NP_033734.3 | 2 - 3 | 207 | 79 | |
| GenBank mRNA | AK047974.1 | - | 2 - 3 | 208 | 79 | |
| JF520813.1 | - | 2 - 3 | 208 | 79 | ||
| JN164666.1 | - | 1 - 2 | 266 | 79 | ||
| M30514.1 | - | 2 - 3 | 248 | 79 | ||
| X03818.1 | - | 2 - 3 | 204 | 79 | ||
| X03819.1 | - | 1 - 2 | 139 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chrng
Genomic Map