Assay Details
| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009604.3 | NP_033734.3 | 4 - 5 | 362 | 88 | |
| GenBank mRNA | AK047974.1 | - | 4 - 5 | 363 | 88 | |
| JN164665.1 | - | 530 | 88 | |||
| JN164666.1 | - | 3 - 4 | 421 | 88 | ||
| M30514.1 | - | 4 - 5 | 403 | 88 | ||
| X03818.1 | - | 4 - 5 | 359 | 88 | ||
| X03819.1 | - | 3 - 4 | 294 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chrng
Genomic Map