Assay Details
| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009604.3 | NP_033734.3 | 8 - 9 | 929 | 76 | |
| GenBank mRNA | AK047974.1 | - | 8 - 9 | 930 | 76 | |
| M30514.1 | - | 8 - 9 | 970 | 76 | ||
| X03818.1 | - | 8 - 9 | 926 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chrng
Genomic Map