Assay Details
| Gene Symbol: Atp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009724.2 | NP_033854.1 | 4 - 5 | 580 | 90 | |
| GenBank mRNA | AK008180.1 | - | 4 - 5 | 579 | 90 | |
| AK009499.1 | - | 4 - 5 | 580 | 90 | ||
| AK009670.1 | - | 4 - 5 | 585 | 90 | ||
| AK009994.1 | - | 4 - 5 | 579 | 90 | ||
| AK010003.1 | - | 4 - 5 | 579 | 90 | ||
| AK010030.1 | - | 4 - 5 | 580 | 90 | ||
| AK010049.1 | - | 4 - 5 | 627 | 90 | ||
| BC120705.1 | - | 4 - 5 | 579 | 40129628 | 90 | |
| BC125292.1 | - | 4 - 5 | 579 | 40129807 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp4b
Genomic Map