Assay Details
| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109757.2 | NP_001103227.1 | 13 - 14 | 2928 | 83 | |
| NM_009726.5 | NP_033856.3 | 13 - 14 | 2925 | 83 | ||
| GenBank mRNA | AB007134.1 | - | 12 - 13 | 2764 | 83 | |
| BC141395.1 | - | 13 - 14 | 2806 | 9056008 | 83 | |
| U03434.1 | - | 13 - 14 | 2833 | 83 | ||
| U03736.1 | - | 12 - 13 | 2673 | 83 | ||
| U71091.1 | - | 13 - 14 | 2829 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp7a
Genomic Map