Assay Details
| Gene Symbol: Bub1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009773.3 | NP_033903.2 | 3 - 4 | 452 | 77 | |
| GenBank mRNA | AF107296.1 | - | 3 - 4 | 407 | 77 | |
| AK133909.1 | - | 3 - 4 | 393 | 77 | ||
| BC031577.1 | - | 3 - 4 | 403 | 4953223 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bub1b
Genomic Map