Assay Details
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009852.3 | NP_033982.3 | 9 - 10 | 1702 | 100 | |
| GenBank mRNA | AK030822.1 | - | 9 - 10 | 1672 | 100 | |
| AK030823.1 | - | 9 - 10 | 1703 | 100 | ||
| AK141213.1 | - | 5 - 6 | 849 | 100 | ||
| BC014294.1 | - | 9 - 10 | 1708 | 4187689 | 100 | |
| U37543.1 | - | 9 - 10 | 1659 | 100 | ||
| U37544.1 | - | 9 - 10 | 1659 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd6
Genomic Map