Assay Details
| Gene Symbol: Cxcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009909.3 | NP_034039.1 | 1 - 2 | 127 | 67 | |
| GenBank mRNA | AK158234.1 | - | 1 - 3 | 133 | 67 | |
| BC051677.1 | - | 1 - 3 | 100 | 30023687 | 67 | |
| CJ056061.1 | - | 1 - 2 | 128 | 67 | ||
| D17630.1 | - | 1 - 2 | 123 | 67 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK158234.1,BC051677.1,D17630.1,NM_009909.3 transcripts.
Set Membership: |
|

Cxcr2
Genomic Map