Assay Details
| Gene Symbol: Cox6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009943.2 | NP_034073.2 | 2 - 3 | 276 | 86 | |
| GenBank mRNA | AK002506.1 | - | 2 - 3 | 227 | 86 | |
| BC028514.1 | - | 2 - 3 | 247 | 1447375 | 86 | |
| BF472154.1 | - | 2 - 3 | 159 | 86 | ||
| BU842968.1 | - | 2 - 3 | 276 | 86 | ||
| U08439.1 | - | 2 - 3 | 262 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | U08439.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | U08439.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | U08439.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | U08439.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | U08439.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | U08439.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25509506 | - | - |
| Whole Mouse Genome G4122A | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | U08439.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | BU842968.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | U08439.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | AK002506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | BC028514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P509997 | BF472154.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122482 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133039 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06588928 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06588925 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05977862 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06588926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25509506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06588928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06632443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05920996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06619682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05905043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05920998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05920992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25509495 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568372 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417607_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457633_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568369 | - | - |
| Mouse Genome 430 2.0 Array | 1417607_at | - | - |
| Mouse Genome 430 2.0 Array | 1457633_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1457633_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1417607_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12862_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12862_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700036594.mm.1 | NM_009943.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036594.mm.1 | NM_009943.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036594.mm.1 | TC0700004358.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036595.mm.1 | NM_009943.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036595.mm.1 | NM_009943.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036595.mm.1 | TC0700004358.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417607_at | - | - |
| Mouse Expression Set 430 | 1417607_PM_at | - | - |
| Mouse Expression Set 430 | 1457633_x_at | - | - |
| Mouse Expression Set 430 | 1457633_PM_x_at | - | - |
| Murine Genome U74v2 Set | 99667_PM_at | - | - |
| Murine Genome U74v2 Set | 99667_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417607_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417607_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417607_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417607_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457633_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457633_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 351154 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 130215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 130215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000130215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2629581 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2629581 | - | - |
| mouse-6 v1.1 (Illumina) | scl30620.3.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 130215 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30620.3.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130215 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2629581 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2629581 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl30620.3.1_5-S | - | - |

Cox6a2
Genomic Map