Assay Details
| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009970.2 | NP_034100.2 | 7 - 8 | 879 | 77 | |
| GenBank mRNA | BC150696.1 | - | 7 - 8 | 819 | 9087607 | 77 |
| EF114317.1 | - | 6 - 7 | 724 | 77 | ||
| EF114318.1 | - | 5 - 6 | 958 | 77 | ||
| M85078.1 | - | 7 - 8 | 820 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229420 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229539 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229409 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | M85078.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | M85078.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288659 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178165 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | M85078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | M85078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | ETG05_66023 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | M85078.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | M85078.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32229421 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178165 | - | - |
| Whole Mouse Genome G4122A | A_51_P341465 | M85078.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341465 | M85078.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341465 | AK135114.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341465 | AW826169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341465 | NM_009970.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178165 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229539 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32229420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32229409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32229444 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468897 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420703_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468893 | - | - |
| Mouse Genome 430 2.0 Array | 1420703_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420703_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12982_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12982_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900015558.mm.1 | NM_009970.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015558.mm.1 | NM_009970.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015558.mm.1 | TC1900001762.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420703_PM_at | - | - |
| Mouse Expression Set 430 | 1420704_at | - | - |
| Mouse Expression Set 430 | 1420704_PM_at | - | - |
| Mouse Expression Set 430 | 1420703_at | - | - |
| Murine Genome U74v2 Set | 99330_PM_at | - | - |
| Murine Genome U74v2 Set | 99330_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420704_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420703_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420704_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420703_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420703_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420703_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 665530 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 520309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520309 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229406 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229406 | - | - |
| mouse-6 v1.1 (Illumina) | scl52308.5.1_95-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52308.5.1_95-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520309 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2866276 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2866276 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl52308.5.1_95-S | - | - |

Csf2ra
Genomic Map