Assay Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291799.1 | NP_001278728.1 | 4 - 5 | 676 | 57 | |
| NM_010070.4 | NP_034200.4 | 4 - 5 | 678 | 57 | ||
| GenBank mRNA | AA790529.1 | - | 2 - 3 | 213 | 57 | |
| AK136382.1 | - | 4 - 5 | 679 | 57 | ||
| BC013066.1 | - | 4 - 5 | 672 | 3498170 | 57 | |
| U78818.2 | - | 4 - 5 | 643 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590979 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590984 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | U78818.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | U78818.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 879697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89594 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | U78818.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | U78818.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P366842 | BC013066.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P366842 | BI135039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P366842 | U78818.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P366842 | NM_010070.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P366842 | AK136382.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | U78818.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | U78818.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24590967 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05960418 | - | - |
| Whole Mouse Genome G4122A | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | U78818.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | BC013066.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | BI135039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | U78818.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | NM_010070.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P275976 | AK136382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366842 | BC013066.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366842 | BI135039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366842 | U78818.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366842 | NM_010070.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366842 | AK136382.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139265 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590979 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590984 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24590981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24590971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24590985 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545626 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417790_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545623 | - | - |
| Mouse Genome 430 2.0 Array | 1417790_at | - | - |
| Mouse Genome 430 2.0 Array | 1417790_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13448_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13448_PM_at | - | - |
| Mouse Expression Set 430 | 1417790_at | - | - |
| Mouse Expression Set 430 | 1417790_PM_at | - | - |
| Murine Genome U74v2 Set | 102896_PM_at | - | - |
| Murine Genome U74v2 Set | 102896_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417790_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417790_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417790_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417790_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 657562 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1050279 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5220273 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2680112 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2680112 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050279 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5220273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002680112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050279 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005220273 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242767 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257069 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2704141 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242767 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2704141 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257069 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001058.1_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001236.1_24-S | - | - |
| mouse-6 v1.1 (Illumina) | scl28809.4_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050279 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2680112 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5220273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001058.1_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28809.4_20-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001236.1_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5220273 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050279 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1242767 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257069 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2704141 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257069 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242767 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2704141 | - | - |

Dok1
Genomic Map