Assay Details
| Gene Symbol: Lefty1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010094.3 | NP_034224.1 | 1 - 2 | 321 | 68 | |
| GenBank mRNA | AJ000082.1 | - | 1 - 2 | 301 | 68 | |
| AK131943.1 | - | 1 - 2 | 323 | 68 | ||
| BC050221.1 | - | 1 - 2 | 284 | 30005856 | 68 | |
| D83921.1 | - | 1 - 2 | 338 | 68 | ||
| Z73151.1 | - | 1 - 2 | 301 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lefty1
Genomic Map