Assay Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276387.1 | NP_001263316.1 | 3 - 4 | 1257 | 83 | |
| NM_010095.6 | NP_034225.1 | 3 - 4 | 1252 | 83 | ||
| GenBank mRNA | AK053013.1 | - | 3 - 4 | 1253 | 83 | |
| AK139262.1 | - | 3 - 4 | 964 | 83 | ||
| BC049188.1 | - | 2 - 3 | 551 | 6312260 | 83 | |
| BC050922.1 | - | 2 - 3 | 774 | 6409737 | 83 | |
| U82441.1 | - | 2 - 3 | 306 | 83 | ||
| U92703.1 | - | 2 - 3 | 657 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ebf2
Genomic Map