Assay Details
| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010121.2 | NP_034251.2 | 2 - 3 | 624 | 101 | |
| GenBank mRNA | AF076681.1 | - | 2 - 3 | 616 | 101 | |
| BC054809.1 | - | 2 - 3 | 636 | 6839221 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak3
Genomic Map