Assay Details
| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010121.2 | NP_034251.2 | 11 - 12 | 2076 | 116 | |
| GenBank mRNA | AF076681.1 | - | 11 - 12 | 2068 | 116 | |
| AK139679.1 | - | 9 - 10 | 1398 | 116 | ||
| BC054809.1 | - | 11 - 12 | 2088 | 6839221 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak3
Genomic Map