Assay Details
| Gene Symbol: Epor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010149.3 | NP_034279.3 | 3 - 4 | 571 | 90 | |
| GenBank mRNA | AK010968.1 | - | 2 - 3 | 279 | 91 | |
| AK139031.1 | - | 3 - 4 | 470 | 90 | ||
| BC003953.1 | - | 3 - 4 | 424 | 90 | ||
| BC046282.1 | - | 3 - 4 | 501 | 6438591 | 90 | |
| J04843.1 | - | 3 - 4 | 453 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epor
Genomic Map