Assay Details
| Gene Symbol: Fgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010203.5 | NP_034333.1 | 1 - 2 | 574 | 67 | |
| GenBank mRNA | AK028694.1 | - | 1 - 2 | 431 | 67 | |
| AK028894.1 | - | 1 - 2 | 576 | 67 | ||
| BC071227.1 | - | 1 - 2 | 547 | 30443152 | 67 | |
| M30643.1 | - | 1 - 2 | 346 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf5
Genomic Map