| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800002384.mm.1 |
NM_001079908.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002384.mm.1 |
NM_001079909.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002384.mm.1 |
NM_010206.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002384.mm.1 |
TC0800000321.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002387.mm.1 |
NM_001079908.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002387.mm.1 |
NM_001079909.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002387.mm.1 |
NM_010206.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002387.mm.1 |
TC0800000321.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800002386.mm.1 |
NM_001079908.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002386.mm.1 |
NM_001079909.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002386.mm.1 |
NM_010206.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800002386.mm.1 |
TC0800000321.mm.1 |
MTA TC |