Assay Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286370.1 | NP_001273299.1 | 8 - 9 | 870 | 116 | |
| NM_010284.3 | NP_034414.2 | 8 - 9 | 1169 | 116 | ||
| GenBank mRNA | AK143886.1 | - | 8 - 9 | 871 | 116 | |
| AK143925.1 | - | 8 - 9 | 967 | 116 | ||
| BC075720.1 | - | 8 - 9 | 1017 | 5044060 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ghr
Genomic Map