Assay Details
| Gene Symbol: Gnrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010323.1 | NP_034453.1 | 1 - 2 | 563 | 69 | |
| GenBank mRNA | BC061250.1 | - | 1 - 2 | 539 | 30249439 | 69 |
| L01119.1 | - | 1 - 2 | 563 | 69 | ||
| L28757.1 | - | 1 - 2 | 518 | 69 | ||
| L28758.1 | - | 1 - 2 | 518 | 69 | ||
| M93108.1 | - | 1 - 2 | 560 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnrhr
Genomic Map