Assay Details
| Gene Symbol: Lpar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010336.2 | NP_034466.2 | 1 - 2 | 73 | 69 | |
| GenBank mRNA | U70622.1 | - | 1 - 2 | 114 | 69 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_010336.2,U70622.1 transcripts.
Set Membership: |
|

Lpar1
Genomic Map