Assay Details
| Gene Symbol: Gsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010344.4 | NP_034474.4 | 3 - 4 | 660 | 89 | |
| GenBank mRNA | AK019177.1 | - | 3 - 4 | 319 | 89 | |
| AK040136.1 | - | 3 - 4 | 369 | 89 | ||
| AK084328.1 | - | 3 - 4 | 290 | 89 | ||
| AK151063.1 | - | 3 - 4 | 306 | 89 | ||
| AK151247.1 | - | 3 - 4 | 306 | 89 | ||
| AK151674.1 | - | 3 - 4 | 218 | 89 | ||
| AK159218.1 | - | 3 - 4 | 309 | 89 | ||
| AK159678.1 | - | 3 - 4 | 332 | 89 | ||
| AK167393.1 | - | 3 - 4 | 291 | 89 | ||
| BC056357.1 | - | 3 - 4 | 660 | 6813295 | 89 | |
| BC057325.1 | - | 3 - 4 | 660 | 6813295 | 89 | |
| X76341.2 | - | 3 - 4 | 358 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gsr
Genomic Map