Assay Details
| Gene Symbol: Hao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010403.2 | NP_034533.1 | 6 - 7 | 991 | 85 | |
| GenBank mRNA | AF104312.1 | - | 6 - 7 | 991 | 85 | |
| AK149565.1 | - | 6 - 7 | 998 | 85 | ||
| BC018587.1 | - | 6 - 7 | 969 | 85 | ||
| BC119535.1 | - | 6 - 7 | 991 | 40056371 | 85 | |
| BC119536.1 | - | 6 - 7 | 991 | 40056372 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hao1
Genomic Map