Assay Details
| Gene Symbol: Hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172117.1 | NP_001165588.1 | 7 - 8 | 903 | 64 | |
| NM_010407.4 | NP_034537.2 | 7 - 8 | 903 | 64 | ||
| GenBank mRNA | AK149736.1 | - | 7 - 8 | 869 | 64 | |
| AK150290.1 | - | 7 - 8 | 877 | 64 | ||
| AK150709.1 | - | 7 - 8 | 886 | 64 | ||
| AK152506.1 | - | 7 - 8 | 877 | 64 | ||
| AK155975.1 | - | 7 - 8 | 816 | 64 | ||
| AK159234.1 | - | 7 - 8 | 836 | 64 | ||
| AK165315.1 | - | 7 - 8 | 868 | 64 | ||
| BC010478.1 | - | 7 - 8 | 899 | 3978518 | 64 | |
| J03023.1 | - | 7 - 8 | 792 | 64 | ||
| Y00487.1 | - | 7 - 8 | 798 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hck
Genomic Map