Assay Details
| Gene Symbol: Hipk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136065.2 | NP_001129537.1 | 11 - 12 | 2643 | 86 | |
| NM_001294143.1 | NP_001281072.1 | 11 - 12 | 2722 | 86 | ||
| NM_001294144.1 | NP_001281073.1 | 11 - 12 | 2719 | 86 | ||
| NM_010433.2 | NP_034563.2 | 11 - 12 | 2803 | 86 | ||
| GenBank mRNA | AF077659.1 | - | 11 - 12 | 2566 | 86 | |
| AF170301.1 | - | 11 - 12 | 2472 | 86 | ||
| AF170302.1 | - | 11 - 12 | 2553 | 86 | ||
| AF208292.1 | - | 11 - 12 | 2556 | 86 | ||
| AF273680.1 | - | 11 - 12 | 2443 | 86 | ||
| AF333791.1 | - | 11 - 12 | 2346 | 86 | ||
| AF333792.1 | - | 11 - 12 | 2430 | 86 | ||
| BC031904.1 | - | 11 - 12 | 2455 | 5368577 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hipk2
Genomic Map