Assay Details
| Gene Symbol: Slc6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010484.2 | NP_034614.2 | 7 - 8 | 1264 | 77 | |
| GenBank mRNA | AF013604.1 | - | 6 - 7 | 1077 | 77 | |
| BC108978.1 | - | 7 - 8 | 1246 | 40047534 | 77 | |
| BC108979.1 | - | 7 - 8 | 1246 | 40047537 | 77 | |
| X66119.1 | - | 6 - 7 | 858 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a4
Genomic Map