Assay Details
| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010508.2 | NP_034638.2 | 1 - 2 | 203 | 102 | |
| GenBank mRNA | AK040184.1 | - | 1 - 2 | 142 | 102 | |
| AK132431.1 | - | 1 - 2 | 135 | 102 | ||
| AK136950.1 | - | 1 - 2 | 180 | 102 | ||
| AK141630.1 | - | 1 - 2 | 139 | 102 | ||
| AK148246.1 | - | 1 - 2 | 145 | 102 | ||
| AK153583.1 | - | 1 - 2 | 136 | 102 | ||
| AK157555.1 | - | 1 - 2 | 170 | 102 | ||
| BC043935.1 | - | 1 - 2 | 154 | 4507009 | 102 | |
| BC052217.1 | - | 1 - 2 | 163 | 30008497 | 102 | |
| BC052429.1 | - | 1 - 2 | 165 | 6836300 | 102 | |
| M89641.1 | - | 1 - 2 | 168 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifnar1
Genomic Map