Assay Details
| Gene Symbol: Ldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113408.1 | NP_001106879.1 | 6 - 7 | 876 | 54 | |
| NM_010697.1 | NP_034827.1 | 6 - 7 | 1061 | 54 | ||
| GenBank mRNA | AB221569.1 | - | 5 - 6 | 418 | 54 | |
| AB250383.1 | - | 5 - 6 | 415 | 54 | ||
| AF030333.1 | - | 6 - 7 | 579 | 54 | ||
| AK048463.1 | - | 1 - 2 | 101 | 54 | ||
| BC013624.1 | - | 6 - 7 | 701 | 3708169 | 54 | |
| U69270.1 | - | 6 - 7 | 1061 | 54 | ||
| U70375.1 | - | 5 - 6 | 411 | 54 | ||
| U89488.1 | - | 6 - 7 | 659 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldb1
Genomic Map