Assay Details
| Gene Symbol: Anxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010730.2 | NP_034860.2 | 10 - 11 | 877 | 86 | |
| GenBank mRNA | AK002681.1 | - | 10 - 11 | 878 | 86 | |
| AK140838.1 | - | 10 - 11 | 880 | 86 | ||
| AK150564.1 | - | 10 - 11 | 879 | 86 | ||
| AK150718.1 | - | 10 - 11 | 879 | 86 | ||
| AK153156.1 | - | 10 - 11 | 879 | 86 | ||
| AK167612.1 | - | 10 - 11 | 879 | 86 | ||
| BC002289.1 | - | 9 - 10 | 832 | 3590168 | 86 | |
| BC004594.1 | - | 9 - 10 | 818 | 3587973 | 86 | |
| CT009771.1 | - | 9 - 10 | 803 | 86 | ||
| CT010298.1 | - | 9 - 10 | 803 | 86 | ||
| EU684130.1 | - | 9 - 10 | 803 | 86 | ||
| M24554.1 | - | 9 - 10 | 788 | 86 | ||
| X07486.1 | - | 9 - 10 | 816 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa1
Genomic Map