Assay Details
| Gene Symbol: Mpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122949.2 | NP_001116421.1 | 7 - 8 | 1204 | 105 | |
| NM_001285496.1 | NP_001272425.1 | 7 - 8 | 1024 | 105 | ||
| NM_001285497.1 | NP_001272426.1 | 7 - 8 | 1204 | 105 | ||
| NM_010823.3 | NP_034953.2 | 7 - 8 | 1180 | 105 | ||
| GenBank mRNA | AF360122.1 | - | 7 - 8 | 961 | 105 | |
| AK041166.1 | - | 7 - 8 | 1152 | 105 | ||
| BC101943.1 | - | 7 - 8 | 1161 | 105 | ||
| BC103513.1 | - | 7 - 8 | 1137 | 105 | ||
| BC103514.1 | - | 7 - 8 | 1161 | 105 | ||
| BC103515.1 | - | 7 - 8 | 1161 | 105 | ||
| BC146612.1 | - | 7 - 8 | 1133 | 105 | ||
| BC146613.1 | - | 7 - 8 | 1138 | 40039147 | 105 | |
| X73677.1 | - | 7 - 8 | 1148 | 105 | ||
| Z22649.1 | - | 7 - 8 | 1138 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpl
Genomic Map