Assay Details
| Gene Symbol: Myl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010861.3 | NP_034991.3 | 5 - 6 | 397 | 88 | |
| GenBank mRNA | AK002367.1 | - | 5 - 6 | 398 | 88 | |
| AK146674.1 | - | 5 - 6 | 401 | 88 | ||
| BC061144.1 | - | 398 | 30301284 | 88 | ||
| M91602.1 | - | 5 - 6 | 389 | 88 | ||
More Information
Set Membership: |
|

Myl2
Genomic Map