Assay Details
| Gene Symbol: Nhlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M82874.1 | - | 1 - 2 | 276 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the M82874.1 transcripts.
Set Membership: |
|

Nhlh1
Genomic Map