Assay Details
| Gene Symbol: Nos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010927.3 | NP_035057.1 | 12 - 13 | 1646 | 82 | |
| GenBank mRNA | AF065919.1 | - | 12 - 13 | 1643 | 82 | |
| AF065920.1 | - | 12 - 13 | 1643 | 82 | ||
| AF065921.2 | - | 12 - 13 | 1643 | 82 | ||
| AF065922.2 | - | 11 - 12 | 1516 | 82 | ||
| AF065923.2 | - | 11 - 12 | 1516 | 82 | ||
| AY090567.1 | - | 11 - 12 | 1466 | 82 | ||
| BC062378.1 | - | 12 - 13 | 1655 | 4978648 | 82 | |
| M84373.1 | - | 12 - 13 | 1647 | 82 | ||
| M87039.1 | - | 12 - 13 | 1718 | 82 | ||
| M92649.1 | - | 12 - 13 | 1654 | 82 | ||
| U43428.1 | - | 12 - 13 | 1534 | 82 | ||
More Information
Set Membership: |
|

Nos2
Genomic Map