Assay Details
| Gene Symbol: Abcb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011076.2 | NP_035206.2 | 9 - 10 | 1118 | 87 | |
| GenBank mRNA | AY864315.1 | - | 8 - 9 | 981 | 87 | |
| M24417.1 | - | 3 - 4 | 467 | 87 | ||
| M30697.1 | - | 9 - 10 | 1132 | 87 | ||
| M33581.1 | - | 9 - 10 | 1117 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1a
Genomic Map