Assay Details
| Gene Symbol: Eif2ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011163.4 | NP_035293.1 | 2 - 3 | 303 | 106 | |
| GenBank mRNA | AK028602.1 | - | 2 - 3 | 303 | 106 | |
| BC016422.1 | - | 2 - 3 | 266 | 3964935 | 106 | |
| M93567.1 | - | 2 - 3 | 245 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2ak2
Genomic Map