Assay Details
| Gene Symbol: Rangap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146174.1 | NP_001139646.1 | 8 - 9 | 1136 | 106 | |
| NM_011241.4 | NP_035371.4 | 9 - 10 | 1184 | 106 | ||
| GenBank mRNA | AK042930.1 | - | 8 - 9 | 1102 | 106 | |
| AK088768.1 | - | 8 - 9 | 1058 | 106 | ||
| AK129452.1 | - | 8 - 9 | 1065 | 106 | ||
| AK133905.1 | - | 8 - 9 | 1104 | 106 | ||
| AK145459.1 | - | 7 - 8 | 739 | 106 | ||
| AK146670.1 | - | 9 - 10 | 1144 | 106 | ||
| AK159009.1 | - | 8 - 9 | 1089 | 106 | ||
| AK159525.1 | - | 8 - 9 | 1101 | 106 | ||
| AK159930.1 | - | 8 - 9 | 1092 | 106 | ||
| AK205795.1 | - | 1 - 2 | 96 | 106 | ||
| BC014855.1 | - | 8 - 9 | 1092 | 3157225 | 106 | |
| BC052862.1 | - | 8 - 9 | 1059 | 30063509 | 106 | |
| BC066213.1 | - | 8 - 9 | 1100 | 30466595 | 106 | |
| BC071200.1 | - | 9 - 10 | 1300 | 5032610 | 106 | |
| U08110.1 | - | 9 - 10 | 1167 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rangap1
Genomic Map