Assay Details
| Gene Symbol: Rangap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146174.1 | NP_001139646.1 | 10 - 11 | 1311 | 186 | |
| NM_011241.4 | NP_035371.4 | 11 - 12 | 1359 | 186 | ||
| GenBank mRNA | AK042930.1 | - | 10 - 11 | 1277 | 186 | |
| AK088768.1 | - | 10 - 11 | 1233 | 186 | ||
| AK129452.1 | - | 10 - 11 | 1240 | 186 | ||
| AK133905.1 | - | 10 - 11 | 1279 | 186 | ||
| AK145459.1 | - | 9 - 10 | 914 | 186 | ||
| AK146670.1 | - | 11 - 12 | 1319 | 186 | ||
| AK159009.1 | - | 10 - 11 | 1264 | 186 | ||
| AK159525.1 | - | 10 - 11 | 1276 | 186 | ||
| AK159930.1 | - | 10 - 11 | 1267 | 186 | ||
| BC014855.1 | - | 10 - 11 | 1267 | 3157225 | 186 | |
| BC052862.1 | - | 10 - 11 | 1234 | 30063509 | 186 | |
| BC066213.1 | - | 10 - 11 | 1275 | 30466595 | 186 | |
| BC071200.1 | - | 11 - 12 | 1475 | 5032610 | 186 | |
| U08110.1 | - | 11 - 12 | 1342 | 186 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rangap1
Genomic Map