Assay Details
| Gene Symbol: Rarg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011244.4 | NP_035374.3 | 1 - 2 | 286 | 128 | |
| GenBank mRNA | AK148064.1 | - | 1 - 2 | 283 | 128 | |
| BC012923.1 | - | 1 - 2 | 212 | 3154037 | 128 | |
| M32068.1 | - | 1 - 2 | 188 | 129 | ||
| M34476.1 | - | 1 - 2 | 43 | 128 | ||
| X15848.1 | - | 1 - 2 | 106 | 128 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK148064.1,BC012923.1,M32068.1,M34476.1,NM_011244.4,X15848.1 transcripts.
Set Membership: |
|

Rarg
Genomic Map